Learn More
Trajan Scientific and Medical™ Replacement Plunger for Spark Holland Syringes
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan Scientific and Medical™ 0318348
Additional Details : Weight : 0.01000kg
Product Code. 15268553
  • 925.00 NOK / Each
Estimated Shipment: 28-06-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement plunger
Gas tight syringes
60 mm
250 μL