missing translation for 'onlineSavingsMsg'
Learn More

RELN Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20689-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890265

  • 1281.92 NOK / 20µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.
Specifications

Specifications

RELN
Polyclonal
Unconjugated
RELN
PRO1598, reelin, RELN, RL
Rabbit
Antigen Affinity Chromatography
RUO
19699, 5649
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P78509, Q60841
RELN
Peptide
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers