missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Xenopus TGF-beta 1 / TGFB1 Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6781-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001081330.1 | |
TGF-beta 1 / TGFB1 Protein | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
Yeast | |
GVGQEYCFGN NGPNCCVKPL YINFRKDLGW KWIHEPKGYE ANYCLGNCPY IWSMDTQYSK VLSLYNQNNP GASISPCCVP DVLEPLPIIY YVGRTAKVEQ LSNMVVRSCN CS | |
Greater than 90% as determined by SDS-PAGE. |
397778 | |
200 μg | |
Not determined. | |
Full Length | |
Xenopus | |
His | |
Tris-based buffer, 50% glycerol |