missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep TGF-alpha Protein

A DNA sequence encoding the Ovis aries (Sheep) TGF-alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6777-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015770

  • 9233.45 NOK / 100µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TGF-alpha Protein
100 μg
Not determined.
Extracellular Domain
Sheep
GST
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TGFA
Recombinant Protein
E. coli
ENSTSALSDP PVAAAVVSHF NDCPDSHTQF CFHGTCRFLL QEEKPACVCH SGYVGARCEH ADLLAVVAAS QKKQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers