missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae SPT15 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) SPT15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6764-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005670

  • 3633.50 NOK / 10µg


Estimated Shipment
16-11-2020
Add to basket

Specifications

Specifications

S. cerevisiae SPT15 Protein
10 μg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SPT15
Recombinant Protein
Yeast
ADEERLKEFK EANKIVFDPN TRQVWENQNR DGTKPATTFQ SEEDIKRAAP ESEKDTSATS GIVPTLQNIV ATVTLGCRLD LKTVALHARN AEYNPKRFAA VIMRIREPKT TALIFASGKM VVTGAKSEDD SKLASRKYAR IIQKIGFAAK FTDFKIQNIV GSCDVKFPIR LEGLAFSHGT FSSYEPELFP GLIYRMVKPK IVLLIFVSGK IVLTGAKQRE EIYQAFEAIY PVLSEFRKM
Videos
Documents