missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PRNP / Prion Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PRNP / Prion, was expressed in the hosts and tags indicated.
2526.25 NOK - 27321.13 NOK
Specifications
Accession Number | NP_036763.1 |
---|---|
Gene ID (Entrez) | 24686 |
Name | PRNP / Prion Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063980
|
enQuireBio™
QP6532-YE-100UG |
100 μg |
7745.00 NOK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003990
|
enQuireBio™
QP6532-YE-500UG |
500 μg |
22140.00 NOK
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073980
|
enQuireBio™
QP6532-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093980
|
enQuireBio™
QP6532-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013990
|
enQuireBio™
QP6532-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083980
|
enQuireBio™
QP6532-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036763.1 | |
PRNP / Prion Protein | |
Not determined. | |
Full Length of Mature Protein | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
24686 | |
Research Use Only | |
Prnp | |
Recombinant Protein | |
Yeast | |
GGWNTGGSRY PGQGSPGGNR YPPQSGGTWG QPHGGGWGQP HGGGWGQPHG GGWGQPHGGG WSQGGGTHNQ WNKPSKPKTN LKHVAGAAAA GAVVGGLGGY MLGSAMSRPM LHFGNDWEDR YYRENMYRYP NQVYYRPVDQ YSNQNNFVHD CVNITIKQHT VTTTTKGENF TETDVKMMER VVEQMCVTQY QKESQAYYDG RRS | |
Greater than 90% as determined by SDS-PAGE. |