missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat MMP7 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) MMP7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6371-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002820

  • 7745.00 NOK / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036996.1
MMP7 Protein
Research Use Only
Mmp7
Recombinant Protein
E. coli
FSLMPNSPKW HSRTVTYRIV SYTTDLPRFL VDQIVKRALR MWSMQIPLNF KRVSWGTADI IIGFARGDHG DNFPFDGPGN TLGHAFAPGP GLGGDAHFDK DEYWTDGEDS GVNFLFVATH ELGHSLGLGH SSVPSSVMYP TYQGDHSEDF SLTKDDIAGI QKLYGKRNKL
Greater than 90% as determined by SDS-PAGE.
25335
100 μg
Not determined.
Full Length
Rat
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers