missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant RABV NucleoProtein

A DNA sequence encoding the Rabies virus (strain PM) (RABV) Nucleoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7060-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017280

  • 11060.00 NOK / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RABV NucleoProtein
Research Use Only
N
Recombinant Protein
Yeast
MDADKIVFKV NNQVVSLKPE IIVDQYEYKY PAIKDLKKPC ITLGKAPDLN KAYKSVLSGM NAAKLDPDDV CSYLAAAMQF FEGTCPEDWT SYGILIARKG DRITPNSLVE IKRTDVEGNW ALTGGMELTR DPTVSEHASL VGLLLSLYRL SKISGQNTGN YKTNIADRIE QIFETAPFVK IVEHHTLMTT HKMCANWSTI PNFRFLAGTY DMFFSRIEHL YSAIRVGTVV TAYEDCSGLV SFTGFIKQIN LTAREAILYF FHKNFEEEIR RMFEPGQETA VPHSYFIHFR SLGLSGKSPY SSNAVGHVFN LIHFVGCYMG QVRSLNATVI AACAPHEMSV LGGYLGEEFF GKGTFERRFF RDEKELQEYE AAELTKTDVA LADDGTVNSD DEDYFSGETR SPEAVYTRIM MNGGRLKRSH IRRYVSVSSN HQARPNSFAE FLNKTYSNDS
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
RABV
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers