missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine Mutant Prolactin Antagonist Protein
A cDNA sequence encoding the Mutant Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14011-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
Mutant Prolactin Antagonist Protein | |
Research Use Only | |
Ovine Prolactin Antagonist mutant form is devoid of agonistic activity and capable of inhibiting biological activity of oPRL or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells. The truncated form is more potent inhibitor. | |
Ovine | |
His | |
Greater than 99.0% as determined by Gel Filtration & SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3. |