missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine Mutant Prolactin Antagonist Protein
A cDNA sequence encoding the Mutant Prolactin Antagonist was constructed and used to recombinantly synthesize the protein.
1380.00 NOK - 20735.00 NOK
Specifications
Name | Mutant Prolactin Antagonist Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Ovine Prolactin Antagonist mutant form is devoid of agonistic activity and capable of inhibiting biological activity of oPRL or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells. The truncated form is more potent inhibitor. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945409
|
enQuireBio™
QP14011-EC-10UG |
10 μg |
1380.00 NOK
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965409
|
enQuireBio™
QP14011-EC-50UG |
50 μg |
2140.00 NOK
50µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955409
|
enQuireBio™
QP14011-EC-1MG |
1 mg |
20735.00 NOK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Mutant Prolactin Antagonist Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
Ovine Prolactin was lyophilized from a concentrated (1 mg/ml) solution with 0.02%-0.03% NaHCO3. |
Research Use Only | |
Ovine Prolactin Antagonist mutant form is devoid of agonistic activity and capable of inhibiting biological activity of oPRL or other lactogenic hormones as evidenced by proliferation assay of Nb2 or other cells. The truncated form is more potent inhibitor. | |
Ovine | |
His | |
Greater than 99.0% as determined by Gel Filtration & SDS-PAGE. |