missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Sex-determining region Y Protein

A DNA sequence encoding the Mus musculus (Mouse) Sex-determining region Y protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6730-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_035694.1
Sex-determining region Y Protein
Research Use Only
Sry
Recombinant Protein
E. coli
MEGHVKRPMN AFMVWSRGER HKLAQQNPSM QNTEISKQLG CRWKSLTEAE KRPFFQEAQR LKILHREKYP NYKYQPHRRA KVSQRSGILQ PAVASTKLYN LLQWDRNPHA ITYRQDWSRA AHLYSKNQQS FYWQPVDIPT GHLQ
Greater than 90% as determined by SDS-PAGE.
21674
50 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers