missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse REG3G / PAP1B Protein

A DNA sequence encoding the Mus musculus (Mouse) REG3G / PAP1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6595-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004420

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_035390.1
REG3G / PAP1B Protein
Research Use Only
Reg3g
Recombinant Protein
Yeast
EVAKKDAPSS RSSCPKGSRA YGSYCYALFS VSKNWYDADM ACQKRPSGHL VSVLSGAEAS FLSSMIKSSG NSGQYVWIGL HDPTLGYEPN RGGWEWSNAD VMNYINWETN PSSSSGNHCG TLSRASGFLK WRENYCNLEL PYVCKFKA
Greater than 90% as determined by SDS-PAGE.
19695
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers