missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein cereblon Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein cereblon, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9830-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067424.2 | |
Protein cereblon Protein | |
Research Use Only | |
Crbn | |
Recombinant Protein | |
Yeast | |
MAGEGDQQDA AHNMGNHLPL LPADSEDEDD EIEMEVEDQD SKEARKPNII NFDTSLPTSH TYLGADMEEF HGRTLHDDDS CQVIPVLPEV LMILIPGQTL PLQLSHPQEV SMVRNLIQKD RTFAVLAYSN VQEREAQFGT TAEIYAYREE QEFGIEVVKV KAIGRQRFKV LELRTQSDGI QQAKVQILPE CVLPSTMSAV QLESLNKCQV FPSKPISWED QYSCKWWQKY QKRKFHCANL TSWPRWLYSL YDAETLMDRI KKQLREWDEN LKDDSLPENP IDFSYRVAAC LPIDDVLRIQ LLKIGSAIQR LRCELDIMNK CTSLCCKQCQ ETEITTKNEI FSLSLCGPMA AYVNPHGYVH ETLTVYKASN LNLIGRPSTV HSWFPGYAWT IAQCKICASH IGWKFTATKK DMSPQKFWGL TRSALLPTIP ETEDEISPDK VILCL | |
Greater than 80% as determined by SDS-PAGE. |
58799 | |
100 μg | |
Not determined. | |
Full. Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |