missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PLBD2 Protein

A DNA sequence encoding the Mus musculus (Mouse) PLBD2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7649-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020011

  • 16028.79 NOK / 100µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_076114.2
PLBD2 Protein
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
71772
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Plbd2
Recombinant Protein
Mammal
LPTLGPGWQR QNPDPPVSRT RSLLLDAASG QLRLEDGFHP DAVAWANLTN AIRETGWAYL DLSTNGRYND SLQAYAAGVV EASVSEELIY MHWMNTVVNY CGPFEYEVGY CEKLKNFLEA NLEWMQREME LNPDSPYWHQ VRLTLLQLKG LEDSYEGRLT FPTGRFTIKP LGFLLLQISG DLEDLEPALN KTNTKPSLGS GSCSALIKLL PGGHDLLVAH NTWNSYQNML RIIKKYRLQF REGPQEEYPL VAGNNLVFSS YPGTIFSGDD FYILGSGLVT LETTIGNKNP ALWKYVQPQG CVLEWIRNVV ANRLALDGAT WADVFKRFNS GTYNNQWMIV DYKAFLPNGP SPGSRVLTIL EQIPGMVVVA DKTAELYKTT YWASYNIPYF ETVFNASGLQ ALVAQYGDWF SYTKNPRAKI FQRDQSLVED MDAMVRLMRY NDFLHDPLSL CEACNPKPNA ENAISARSDL NPANGSYPFQ ALHQRAHGGI DVKVTSFTLA KYMSMLAASG PTWDQCPPFQ WSKSPFHSML HMGQPDLWMF SPIRVPWD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers