missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Integrin alpha-L Protein

A DNA sequence encoding the Mus musculus (Mouse) Integrin alpha-L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6240-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011860

  • 20726.60 NOK / 500µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Integrin alpha-L Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Itgal
Recombinant Protein
Yeast
DLVFLFDGSQ SLDRKDFEKI LEFMKDVMRK LSNTSYQFAA VQFSTDCRTE FTFLDYVKQN KNPDVLLGSV QPMFLLTNTF RAINYVVAHV FKEESGARPD ATKVLVIITD GEASDKGNIS AAHDITRYII GIGKHFVSVQ KQKTLHIFAS EPVEEFVKIL DTFEKLKDLF TDL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers