missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL1F10 / IL-38 Protein

A DNA sequence encoding the Mus musculus (Mouse) IL1F10 / IL-38, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7885-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011401

  • 20726.60 NOK / 500µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_694717.1
IL1F10 / IL-38 Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
215274
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Il1f10
Recombinant Protein
Yeast
MCSLPMARYY IIKDAHQKAL YTRNGQLLLG DPDSDNYSPE KVCILPNRGL DRSKVPIFLG MQGGSCCLAC VKTREGPLLQ LEDVNIEDLY KGGEQTTRFT FFQRSLGSAF RLEAAACPGW FLCGPAEPQQ PVQLTKESEP STHTEFYFEM SR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers