missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Heme oxygenase 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Heme oxygenase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9180-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017791

  • 20726.60 NOK / 500µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_034572.1
Heme oxygenase 1 Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
15368
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Hmox1
Recombinant Protein
Yeast
MERPQPDSMP QDLSEALKEA TKEVHIQAEN AEFMKNFQKG QVSREGFKLV MASLYHIYTA LEEEIERNKQ NPVYAPLYFP EELHRRAALE QDMAFWYGPH WQEIIPCTPA TQHYVKRLHE VGRTHPELLV AHAYTRYLGD LSGGQVLKKI AQKAMALPSS GEGLAFFTFP NIDSPTKFKQ LYRARMNTLE MTPEVKHRVT EEAKTAFLLN IELFEELQVM LTEEHKDQSP SQMASLRQRP ASLVQDTAPA ETPRGKPQIS TSSSQTPLLQ WVLTLSFLLA TVAVGIYAM
Videos
SDS
Documents
Certificates
Special Offers

Special Offers