missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Hadh, mitochondrial Protein

A DNA sequence encoding the Mus musculus (Mouse) Hadh, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7685-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020181

  • 32203.54 NOK / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032238.2
Hadh, mitochondrial Protein
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
15107
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Hadh
Recombinant Protein
E. coli
SSSSSASAAA KKILIKHVTV IGGGLMGAGI AQVAAATGHT VVLVDQTEDI LAKSKKGIEE SLKRMAKKKF TENPKAGDEF VEKTLSCLST STDAASVVHS TDLVVEAIVE NLKLKNELFQ RLDKFAAEHT IFASNTSSLQ ITNIANATTR QDRFAGLHFF NPVPMMKLVE VIKTPMTSQK TFESLVDFCK TLGKHPVSCK DTPGFIVNRL LVPYLIEAVR LHERGDASKE DIDTAMKLGA GYPMGPFELL DYVGLDTTKF ILDGWHEMEP ENPLFQPSPS MNNLVAQKKL GKKTGEGFYK YK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers