missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fibroblast growth factor 15 Protein
A DNA sequence encoding the Mus musculus (Mouse) Fibroblast growth factor 15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7474-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_032029.1 | |
Fibroblast growth factor 15 Protein | |
Research Use Only | |
Fgf15 | |
Recombinant Protein | |
E. coli | |
RPLAQQSQSV SDEDPLFLYG WGKITRLQYL YSAGPYVSNC FLRIRSDGSV DCEEDQNERN LLEFRAVALK TIAIKDVSSV RYLCMSADGK IYGLIRYSEE DCTFREEMDC LGYNQYRSMK HHLHIIFIQA KPREQLQDQK PSNFIPVFHR SFFETGDQLR SKMFSLPLES DSMDPFRMVE DVDHLVKSPS FQK | |
Greater than 90% as determined by SDS-PAGE. |
14170 | |
1 mg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |