missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Erythroferrone Protein

A DNA sequence encoding the Mus musculus (Mouse) Erythroferrone, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9819-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008781

  • 7144.40 NOK / 100µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NC_000002.12
Erythroferrone Protein
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
151176
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ERFE
Recombinant Protein
Yeast
LGVPESAEPV GTHARPQPPG AELPAPPANS PPEPTIAHAH SVDPRDAWML FVKQSDKGIN SKRRSKARRL KLGLPGPPGP PGPQGPPGPF IPSEVLLKEF QLLLKGAVRQ RESHLEHCTR DLTTPASGSP SRVPAAQELD SQDPGALLAL LAATLAQGPR APRVEAAFHC RLRRDVQVDR RALHELGIYY LPEVEGAFHR GPGLNLTSGQ YTAPVAGFYA LAATLHVALT EQPRKGPTRP RDRLRLLICI QSLCQHNASL ETVMGLENSS ELFTISVNGV LYLQAGHYTS VFLDNASGSS LTVRSGSHFS AILLGL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers