missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EF-hand domain-containing protein D2 Protein
A DNA sequence encoding the Mus musculus (Mouse) EF-hand domain-containing protein D2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8046-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
EF-hand domain-containing protein D2 Protein | |
Research Use Only | |
Efhd2 | |
Recombinant Protein | |
E. coli | |
ATDELASKLS RRLQMEGEGG EATEQPGLNG AAAAAAAEAP DETAQALGSA DDELSAKLLR RADLNQGIGE PQSPSRRVFN PYTEFKEFSR KQIKDMEKMF KQYDAGRDGF IDLMELKLMM EKLGAPQTHL GLKSMIQEVD EDFDSKLSFR EFLLIFRKAA AGELQEDSGL QVLARLSEID VSTEGVKGAK NFFEAKVQAI NVSSRFEEEI KAEQEERKKQ AEEVKQRKAA FKELQSTFK | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |