Learn More
enQuireBio™ Recombinant Mouse Cytochrome P450 2E1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Cytochrome P450 2E1, was expressed in the hosts and tags indicated.
2526.25 NOK - 34365.00 NOK
Specifications
Accession Number | NP_067257.1 |
---|---|
Gene ID (Entrez) | 13106 |
Name | Cytochrome P450 2E1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087631
|
enQuireBio™
QP9094-YE-100UG |
100 μg |
7745.00 NOK
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007641
|
enQuireBio™
QP9094-YE-1MG |
1 mg |
34365.00 NOK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16097631
|
enQuireBio™
QP9094-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017641
|
enQuireBio™
QP9094-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027641
|
enQuireBio™
QP9094-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037641
|
enQuireBio™
QP9094-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_067257.1 | |
Cytochrome P450 2E1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
13106 | |
Research Use Only | |
Cyp2e1 | |
Recombinant Protein | |
Yeast | |
AVLGITVALL VWIATLLLVS IWKQIYRSWN LPPGPFPIPF FGNIFQLDLK DIPKSLTKLA KRFGPVFTLH LGQRRIVVLH GYKAVKEVLL NHKNEFSGRG DIPVFQEYKN KGIIFNNGPT WKDVRRFSLS ILRDWGMGKQ GNEARIQREA HFLVEELKKT KGQPFDPTFL IGCAPCNVIA DILFNKRFDY DDKKCLELMS LFNENFYLLS TPWIQAYNYF SDYLQYLPGS HRKVMKNVSE IRQYTLGKAK EHLKSLDINC PRDVTDCLLI EMEKEKHSQE PMYTMENISV TLADLFFAGT ETTSTTLRYG LLILMKYPEI EEKLHEEIDR VIGPSRAPAV RDRMNMPYMD AVVHEIQRFI NLVPSNLPHE ATRDTVFRGY VIPKGTVVIP TLDSLLFDNY EFPDPETFKP EHFLNENGKF KYSDYFKAFS AGKRVCVGEG LARMELFLLL SAILQHFNLK SLVDPKDIDL SPVTIGFGSI PREFKLCVIP RS | |
Greater than 90% as determined by SDS-PAGE. |