missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cd163 Protein

A DNA sequence encoding the Mus musculus (Mouse) Cd163, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7641-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009970

  • 7287.29 NOK / 100µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_444324.2
Cd163 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
93671
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cd163
Recombinant Protein
E. coli
VVCQQLGCPT SIKALGWANS SAGSGYIWMD KVSCTGNESA LWDCKHDGWG KHNCTHEKDA GVTCSDGSNL EMRLVNSAGH RCLGRVEIKF QGKWGTVCDD NFSKDHASVI CKQLGCGSAI SFSGSAKLGA GSGPIWLDDL ACNGNESALW DCKHRGWGKH NCDHAEDVGV ICLEGADLSL RLVDGVSRCS GRLEVRFQGE WGTVCDDNWD LRDASVVCKQ LGCPTAISAI GRVNASEGSG QIWLDNISCE GHEATLWECK HQEWGKHYCH HREDAGVTCS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers