missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Adiponectin Protein
A DNA sequence encoding the Mus musculus (Mouse) Adiponectin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8593-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033735.3 | |
Adiponectin Protein | |
Research Use Only | |
ma/Adipoq | |
Recombinant Protein | |
E. coli | |
EDDVTTTEEL APALVPPPKG TCAGWMAGIP GHPGHNGTPG RDGRDGTPGE KGEKGDAGLL GPKGETGDVG MTGAEGPRGF PGTPGRKGEP GEAAYVYRSA FSVGLETRVT VPNVPIRFTK IFYNQQNHYD GSTGKFYCNI PGLYYFSYHI TVYMKDVKVS LFKKDKAVLF TYDQYQEKNV DQASGSVLLH LEVGDQVWLQ VYGDGDHNGL YADNVNDSTF TGFLLYHDTN | |
Greater than 90% as determined by SDS-PAGE. |
11450 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |