missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Bangkok/1/1979 H3N2) Influenza A H3N2 (strain A/Bangkok/1/1979) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9635-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H3N2 (Bangkok / 1 / 1979) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
Yeast | |
GIFGAIAGFI ENGWEGMXXG WYGFRHQNSE GTGQAADLKS TQAAIDQING KLNRVIEKTN EKFHQIEKEF SEVEGRIQDL EKYVEDTKID LWSYNAELLV ALENQHTIDL TDSEMNKLFE KTRRQLRENA EDMGNGCFKI YHKCDNACIG SIRNGTYDHD VYRDEALNNR FQIKGVELKX GYKDWILWIS FAISCFLLCV VLLGFIMVXC QKGNIRCNIC I | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |