missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WDR45 Protein
A DNA sequence encoding the Homo sapiens (Human) WDR45, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7978-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001025067.1 | |
WDR45 Protein | |
Research Use Only | |
WDR45 | |
Recombinant Protein | |
E. coli | |
MTQQPLRGVT SLRFNQDQSC FCCAMETGVR IYNVEPLMEK GHLDHEQVGS MGLVEMLHRS NLLALVGGGS SPKFSEISVL IWDDAREGKD SKEKLVLEFT FTKPVLSVRM RHDKIVIVLK NRIYVYSFPD NPRKLFEFDT RDNPKGLCDL CPSLEKQLLV FPGHKCGSLQ LVDLASTKPG TSSAPFTINA HQSDIACVSL NQPGTVVASA SQKGTLIRLF DTQSKEKLVE LRRGTDPATL YCINFSHDSS FLCASSDKGT VHIFALKDTR LNRRSALARV GKVGPMIGQY VDSQWSLASF TVPAESACIC AFGRNTSKNV NSVIAICVDG TFHKYVFTPD GNCNREAFDV YLDICDDDDF | |
Greater than 80% as determined by SDS-PAGE. |
11152 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |