missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VEGFR2 / Flk-1 / CD309 / KDR Protein

A DNA sequence encoding the Homo sapiens (Human) VEGFR2 / Flk-1 / CD309 / KDR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6258-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011990

  • 22559.24 NOK / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002244.1
VEGFR2 / Flk-1 / CD309 / KDR Protein
1 mg
Not determined.
Extracellular Domain
Human
GST
Tris-based buffer, 50% glycerol
3791
Greater than 90% as determined by SDS-PAGE.
Research Use Only
KDR
Recombinant Protein
E. coli
ASVGLPSVSL DLPRLSIQKD ILTIKANTTL QITCRGQRDL DWLWPNNQSG SEQRVEVTEC SDGLFCKTLT IPKVIGNDTG AYKCFYRETD LASVIYVYVQ DYRSPFIASV SDQHGVVYIT ENKNKTVVIP CLGSISNLNV SLCARYPEKR FVPDGNRISW DSKKGFTIPS YMISYAGMVF CEAKINDESY QSIMYIVVVV GYRIYDVVLS PSHGIELSVG EKLVLNCTAR TELNVGIDFN WEYPSSKHQH KKLVNRDLKT QSGSEMKKFL STLTIDGVTR SDQGLYTCAA SSGLMTKKNS TFVRVHEKPF VAFGSGMESL VEATVGERVR IPAKYLGYPP PEIKWYKNGI PLESNHTIKA GHVLTIMEVS ERDTGNYTVI LTNPISKEKQ SHVVSLVVYV PPQIGEKSLI SPVDSYQYGT TQTLTCTVYA IPPPHHIHWY WQLEEECANE PSQAVSVTNP YPCEEWRSVE DFQGGNKIEV NKNQFALIEG KNKTVSTLVI QAANVSALYK CEAVNKVGRG ERVISFHVTR GPEITLQPDM QPTEQESVSL WCTADRSTFE NLTWYKLGPQ PLPIHVGELP TPVCKNLDTL WKLNATMFSN STNDILIMEL KNASLQDQGD YVCLAQDRKT KKRHCVVRQL TVLERVAPTI TGNLENQTTS IGESIEVSCT ASGNPPPQIM WFKDNETLVE DSGIVLKDGN RNLTIRRVRK EDEGLYTCQA CSVLGCAKVE AFFIIEGAQE KTNLE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers