missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSLP Protein

A DNA sequence encoding the Homo sapiens (Human) TSLP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6845-ma-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026250

  • 5719.55 NOK / 20µg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_149024.1
TSLP Protein
20 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
85480
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TSLP
Recombinant Protein
Mammal
YDFTNCDFEK IKAAYLSTIS KDLITYMSGT KSTEFNNTVS CSNRPHCLTE IQSLTFNPTA GCASLAKEMF AMKTKAALAI WCPGYSETQI NATQAMKKRR KRKVTTNKCL EQVSQLQGLW RRFNRPLLKQ Q
Videos
SDS
Documents
Certificates
Special Offers

Special Offers