missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription intermediary factor 1-beta Protein

A DNA sequence encoding the Homo sapiens (Human) Transcription intermediary factor 1-beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8501-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004981

  • 12152.88 NOK / 500µg


Estimated Shipment
24-11-2020
Add to basket

Specifications

Specifications

NP_005753.1
Transcription intermediary factor 1-beta Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
10155
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TRIM28
Recombinant Protein
E. coli
PGEGSAGGEK RSTAPSAAAS ASASAAASSP AGGGAEALEL LEHCGVCRER LRPEREPRLL PCLHSACSAC LGPAAPAAAN SSGDGGAAGD GTVVDCPVCK QQCFSKDIVE NYFMRDSGSK AATDAQDANQ CCTSCEDNAP ATSYCVECSE PLCETCVEAH QRVKYTKDHT VRSTGPAKSR DGERTVYCNV HKHEPLVLFC ESCDTLTCRD CQLNAHKDHQ YQFLEDAVRN QRKLLASLVK RLGDKHATLQ KSTKEVRSSI RQVSDVQKRV
Videos
SDS
Documents