Learn More
enQuireBio™ Recombinant Human Transcription intermediary factor 1-beta Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription intermediary factor 1-beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8501-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_005753.1 | |
Transcription intermediary factor 1-beta Protein | |
Research Use Only | |
TRIM28 | |
Recombinant Protein | |
E. coli | |
PGEGSAGGEK RSTAPSAAAS ASASAAASSP AGGGAEALEL LEHCGVCRER LRPEREPRLL PCLHSACSAC LGPAAPAAAN SSGDGGAAGD GTVVDCPVCK QQCFSKDIVE NYFMRDSGSK AATDAQDANQ CCTSCEDNAP ATSYCVECSE PLCETCVEAH QRVKYTKDHT VRSTGPAKSR DGERTVYCNV HKHEPLVLFC ESCDTLTCRD CQLNAHKDHQ YQFLEDAVRN QRKLLASLVK RLGDKHATLQ KSTKEVRSSI RQVSDVQKRV | |
Greater than 90% as determined by SDS-PAGE. |
10155 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |