missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription elongation factor SPT4 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcription elongation factor SPT4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6753-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015580

  • 32203.54 NOK / 1mg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003159.1
Transcription elongation factor SPT4 Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6827
Greater than 80% as determined by SDS-PAGE.
Research Use Only
SUPT4H1
Recombinant Protein
E. coli
ALETVPKDLR HLRACLLCSL VKTIDQFEYD GCDNCDAYLQ MKGNREMVYD CTSSSFDGII AMMSPEDSWV SKWQRVSNFK PGVYAVSVTG RLPQGIVREL KSRGVAYKSR DTAIKT
Videos
SDS
Documents
Certificates
Special Offers

Special Offers