missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription elongation factor SPT4 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcription elongation factor SPT4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6753-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005580

  • 2526.25 NOK / 10µg


Estimated Shipment
16-11-2020
Add to basket

Specifications

Specifications

NP_003159.1
Transcription elongation factor SPT4 Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6827
Greater than 80% as determined by SDS-PAGE.
Research Use Only
SUPT4H1
Recombinant Protein
E. coli
ALETVPKDLR HLRACLLCSL VKTIDQFEYD GCDNCDAYLQ MKGNREMVYD CTSSSFDGII AMMSPEDSWV SKWQRVSNFK PGVYAVSVTG RLPQGIVREL KSRGVAYKSR DTAIKT
Videos
Documents