missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAR DNA-binding protein 43 Protein

A DNA sequence encoding the Homo sapiens (Human) TAR DNA-binding protein 43, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6761-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005640

  • 1644.75 NOK / 10µg


Estimated Shipment
13-11-2020
Add to basket

Specifications

Specifications

NP_031401.1
TAR DNA-binding protein 43 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
23435
Greater than 80% as determined by SDS-PAGE.
Research Use Only
TARDBP
Recombinant Protein
E. coli
MSEYIRVTED ENDEPIEIPS EDDGTVLLST VTAQFPGACG LRYRNPVSQC MRGVRLVEGI LHAPDAGWGN LVYVVNYPKD NKRKMDETDA SSAVKVKRAV QKTSDLIVLG LPWKTTEQDL KEYFSTFGEV LMVQVKKDLK TGHSKGFGFV RFTEYETQVK VMSQRHMIDG RWCDCKLPNS KQSQDEPLRS RKVFVGRCTE DMTEDELREF FSQYGDVMDV FIPKPFRAFA FVTFADDQIA QSLCGEDLII KGISVHISNA EPKHNSNRQL ERSGRFGGNP GGFGNQGGFG NSRGGGAGLG NNQGSNMGGG MNFGAFSINP AMMAAAQAAL QSSWGMMGML ASQQNQSGPS GNNQNQGNMQ REPNQAFGSG NNSYSGSNSG AAIGWGSASN AGSGSG
Videos
Documents