missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Syntaxin-10 Protein

A DNA sequence encoding the Homo sapiens (Human) Syntaxin-10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6741-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001258540.1
Syntaxin-10 Protein
Research Use Only
STX10
Recombinant Protein
E. coli
SLEDPFFVVR GEVQKAVNTA RGLYQRWCEL LQESAAVGRE ELDWTTNELR NGLRSIEWDL EDLEETIGIV EANPGKFKLP AGDLQERKVF VERMREAVQE MKDHMVSPTA VAFLERNNRE ILAGKPAAQK SPSDLLDASA VSATSRYIEE QQATQQLIMD EQDQQLEMVS GSIQVLKHMS GRVGEELDEQ GIMLDAFAQE MDHTQSRMDG VLRKLAKVSH MTSDRRQ
Greater than 90% as determined by SDS-PAGE.
8677
200 μg
Not determined.
Cytoplasmic Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers