missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Synaptogyrin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Synaptogyrin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6755-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004702.2 | |
Synaptogyrin-1 Protein | |
Research Use Only | |
SYNGR1 | |
Recombinant Protein | |
E. coli | |
MEGGAYGAGK AGGAFDPYTL VRQPHTILRV VSWLFSIVVF GSIVNEGYLN SASEGEEFCI YNRNPNACSY GVAVGVLAFL TCLLYLALDV YFPQISSVKD RKKAVLSDIG VSAFWAFLWF VGFCYLANQW QVSKPKDNPL NEGTDAARAA IAFSFFSIFT WSLTAALAVR RFKDLSFQEE YSTLFPASAQ P | |
Greater than 80% as determined by SDS-PAGE. |
9145 | |
200 μg | |
Not determined. | |
Full Length of Isoform 1B | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |