missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / threonine-protein kinase 25 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase 25, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6739-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001258906.1 | |
Serine / threonine-protein kinase 25 Protein | |
Research Use Only | |
STK25 | |
Recombinant Protein | |
E. coli | |
MAHLRGFANQ HSRVDPEELF TKLDRIGKGS FGEVYKGIDN HTKEVVAIKI IDLEEAEDEI EDIQQEITVL SQCDSPYITR YFGSYLKSTK LWIIMEYLGG GSALDLLKPG PLEETYIATI LREILKGLDY LHSERKIHRD IKAANVLLSE QGDVKLADFG VAGQLTDTQI KRNTFVGTPF WMAPEVIKQS AYDFKADIWS LGITAIELAK GEPPNSDLHP MRVLFLIPKN SPPTLEGQHS KPFKEFVEAC LNKDPRFRPT AKELLKHKFI TRYTKKTSFL TELIDRYKRW KSEGHGEESS SEDSDIDGEA EDGEQGPIWT FPPTIRPSPH SKLHKGTALH SSQKPAEPVK RQPRSQCLST LVRPVFGELK EKHKQSGGSV GALEELENAF SLAEESCPGI SDKLMVHLVE RVQRFSHNRN HLTSTR | |
Greater than 90% as determined by SDS-PAGE. |
10494 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |