missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Selenide, water dikinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Selenide, water dikinase 1, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_001182531.1 |
---|---|
Gene ID (Entrez) | 22929 |
Name | Selenide, water dikinase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
16004910
|
enQuireBio™
QP6663-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024910
|
enQuireBio™
QP6663-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014910
|
enQuireBio™
QP6663-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034910
|
enQuireBio™
QP6663-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044910
|
enQuireBio™
QP6663-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054910
|
enQuireBio™
QP6663-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_001182531.1 | |
Selenide, water dikinase 1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
22929 | |
Research Use Only | |
SEPHS1 | |
Recombinant Protein | |
E. coli | |
STRESFNPES YELDKSFRLT RFTELKGTGC KVPQDVLQKL LESLQENHFQ EDEQFLGAVM PRLGIGMDTC VIPLRHGGLS LVQTTDYIYP IVDDPYMMGR IACANVLSDL YAMGVTECDN MLMLLGVSNK MTDRERDKVM PLIIQGFKDA AEEAGTSVTG GQTVLNPWIV LGGVATTVCQ PNEFIMPDNA VPGDVLVLTK PLGTQVAVAV HQWLDIPEKW NKIKLVVTQE DVELAYQEAM MNMARLNRTA AGLMHTFNAH AATDITGFGI LGHAQNLAKQ QRNEVSFVIH NLPVLAKMAA VSKACGNMFG LMHGTCPETS GGLLICLPRE QAARFCAEIK SPKYGEGHQA WIIGIVEKGN RTARIIDKPR IIEVAPQVAT QNVNPTPGAT S | |
Greater than 80% as determined by SDS-PAGE. |