missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S-phase kinase-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) S-phase kinase-associated protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6689-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005140

  • 2418.75 NOK / 50µg


Estimated Shipment
19-11-2020
Add to basket

Specifications

Specifications

NP_008861.2
S-phase kinase-associated protein 1
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6500
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SKP1
Recombinant Protein
E. coli
PSIKLQSSDG EIFEVDVEIA KQSVTIKTML EDLGMDDEGD DDPVPLPNVN AAILKKVIQW CTHHKDDPPP PEDDENKEKR TDDIPVWDQE FLKVDQGTLF ELILAANYLD IKGLLDVTCK TVANMIKGKT PEEIRKTFNI KNDFTEEEEA QVRKENQWCE EK
Videos
Documents