missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ROR1 Protein

A DNA sequence encoding the Homo sapiens (Human) ROR1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6615-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004570

  • 19129.63 NOK / 1mg


Estimated Shipment
19-11-2020
Add to basket

Specifications

Specifications

NP_001077061.1
ROR1 Protein
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
4919
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ROR1
Recombinant Protein
E. coli
QETELSVSAE LVPTSSWNIS SELNKDSYLT LDEPMNNITT SLGQTAELHC KVSGNPPPTI RWFKNDAPVV QEPRRLSFRS TIYGSRLRIR NLDTTDTGYF QCVATNGKEV VSSTGVLFVK FGPPPTASPG YSDEYEEDGF CQPYRGIACA RFIGNRTVYM ESLHMQGEIE NQITAAFTMI GTSSHLSDKC SQFAIPSLCH YAFPYCDETS SVPKPRDLCR DECEILENVL CQTEYIFARS NPMILMRLKL PNCEDLPQPE SPEAANCIRI GIPMADPINK NHKCYNSTGV DYRGTVSVTK SGRQCQPWNS QYPHTHTFTA LRFPELNGGH SYCRNPGNQK EAPWCFTLDE NFKSDLCDIP AC
Videos
Documents