missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein Wnt-10a Protein

A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-10a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9917-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009001

  • 17555.00 NOK / 500µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_079492.2
Protein Wnt-10a Protein
Research Use Only
WNT10A
Recombinant Protein
Yeast
MPRSAPNDIL DLRLPPEPVL NANTVCLTLP GLSRRQMEVC VRHPDVAASA IQGIQIAIHE CQHQFRDQRW NCSSLETRNK IPYESPIFSR GFRESAFAYA IAAAGVVHAV SNACALGKLK ACGCDASRRG DEEAFRRKLH RLQLDALQRG KGLSHGVPEH PALPTASPGL QDSWEWGGCS PDMGFGERFS KDFLDSREPH RDIHARMRLH NNRVGRQAVM ENMRRKCKCH GTSGSCQLKT CWQVTPEFRT VGALLRSRFH RATLIRPHNR NGGQLEPGPA GAPSPAPGAP GPRRRASPAD LVYFEKSPDF CEREPRLDSA GTVGRLCNKS SAGSDGCGSM CCGRGHNILR QTRSERCHCR FHWCCFVVCE ECRITEWVSV CK
Greater than 90% as determined by SDS-PAGE.
80326
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers