missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Prohibitin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Prohibitin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8027-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002241

  • 4941.08 NOK / 100µg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138303.1
Prohibitin-2 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
11331
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PHB2
Recombinant Protein
E. coli
AQNLKDLAGR LPAGPRGMGT ALKLLLGAGA VAYGVRESVF TVEGGHRAIF FNRIGGVQQD TILAEGLHFR IPWFQYPIIY DIRARPRKIS SPTGSKDLQM VNISLRVLSR PNAQELPSMY QRLGLDYEER VLPSIVNEVL KSVVAKFNAS QLITQRAQVS LLIRRELTER AKDFSLILDD VAITELSFSR EYTAAVEAKQ VAQQEAQRAQ FLVEKAKQEQ RQKIVQAEGE AEAAKMLGEA LSKNPGYIKL RKIRAAQNIS KTIATSQNRI YLTADNLVLN LQDESFTRGS DSLIKGKK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers