missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKCA-binding Protein
A DNA sequence encoding the Homo sapiens (Human) PRKCA-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8506-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034672.1 | |
PRKCA-binding Protein | |
Research Use Only | |
PICK1 | |
Recombinant Protein | |
E. coli | |
MFADLDYDIE EDKLGIPTVP GKVTLQKDAQ NLIGISIGGG AQYCPCLYIV QVFDNTPAAL DGTVAAGDEI TGVNGRSIKG KTKVEVAKMI QEVKGEVTIH YNKLQADPKQ GMSLDIVLKK VKHRLVENMS SGTADALGLS RAILCNDGLV KRLEELERTA ELYKGMTEHT KNLLRAFYEL SQTHRAFGDV FSVIGVREPQ | |
Greater than 90% as determined by SDS-PAGE. |
9463 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |