Learn More
enQuireBio™ Recombinant Human Prestin Protein
A DNA sequence encoding the Homo sapiens (Human) Prestin, was expressed in the hosts and tags indicated.
2085.50 NOK - 22225.63 NOK
Specifications
Accession Number | NP_001161434.1 |
---|---|
Gene ID (Entrez) | 375611 |
Name | Prestin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008251
|
enQuireBio™
QP9401-YE-100UG |
100 μg |
6475.00 NOK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048251
|
enQuireBio™
QP9401-YE-500UG |
500 μg |
17555.00 NOK
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018251
|
enQuireBio™
QP9401-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038251
|
enQuireBio™
QP9401-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058251
|
enQuireBio™
QP9401-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028251
|
enQuireBio™
QP9401-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001161434.1 | |
Prestin Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
375611 | |
Research Use Only | |
SLC26A5 | |
Recombinant Protein | |
Yeast | |
YRTQSPSYKV LGKLPETDVY IDIDAYEEVK EIPGIKIFQI NAPIYYANSD LYSNALKRKT GVNPAVIMGA RRKAMRKYAK EVGNANMANA TVVKADAEVD GEDATKPEEE DGEVKYPPIV IKSTFPEEMQ RFMPPGDNVH TVILDFTQVN FIDSVGVKTL AGIVKEYGDV GIYVYLAGCS AQVVNDLTRN RFFENPALWE LLFHSIHDAV LGSQLREALA EQEASAPPSQ EDLEPNATPA TPEA | |
Greater than 90% as determined by SDS-PAGE. |