missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLTP Protein
A DNA sequence encoding the Homo sapiens (Human) PLTP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6507-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001229849.1 | |
PLTP Protein | |
Research Use Only | |
PLTP | |
Recombinant Protein | |
E. coli | |
EFPGCKIRVT SKALELVKQE GLRFLEQELE TITIPDLRGK EGHFYYNISE VKVTELQLTS SELDFQPQQE LMLQITNASL GLRFRRQLLY WFFYDGGYIN ASAEGVSIRT GLELSRDPAG RMKVSNVSCQ ASVSRMHAAF GGTFKKVYDF LSTFITSGMR FLLNQQICPV LYHAGTVLLN SLLDTVPVRS SVDELVGIDY SLMKDPVAST SNLDMDFRGA FFPLTERNWS LPNRAVEPQL QEEERMVYVA FSEFFFDSAM ESYFRAGALQ LLLVGDKVPH DLDMLLRATY FGSIVLLSPA VIDSPLKLEL RVLAPPRCTI KPSGTTISVT ASVTIALVPP DQPEVQLSSM TMDARLSAKM ALRGKALRTQ LDLRRFRIYS NHSALESLAL IPLQAPLKTM LQIGVMPMLN ERTWRGVQIP LPEGINFVHE VVTNHAGFLT IGADLHFAKG LREVIEKNRP ADVRASTAPT PSTAAV | |
Greater than 90% as determined by SDS-PAGE. |
5360 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |