missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phosphomannomutase 2 / PMM2 / CDG1 Protein
A DNA sequence encoding the Homo sapiens (Human) Phosphomannomutase 2 / PMM2 / CDG1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6508-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000294.1 | |
Phosphomannomutase 2 / PMM2 / CDG1 Protein | |
Research Use Only | |
PMM2 | |
Recombinant Protein | |
E. coli | |
AAPGPALCLF DVDGTLTAPR QKITKEMDDF LQKLRQKIKI GVVGGSDFEK VQEQLGNDVV EKYDYVFPEN GLVAYKDGKL LCRQNIQSHL GEALIQDLIN YCLSYIAKIK LPKKRGTFIE FRNGMLNVSP IGRSCSQEER IEFYELDKKE NIRQKFVADL RKEFAGKGLT FSIGGQISFD VFPDGWDKRY CLRHVENDGY KTIYFFGDKT MPGGNDHEIF TDPRTMGYSV TAPEDTRRIC ELLFS | |
Greater than 80% as determined by SDS-PAGE. |
5373 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |