missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDZD11 / PDZK11 / PISP Protein

A DNA sequence encoding the Homo sapiens (Human) PDZD11 / PDZK11 / PISP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7680-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16010141

  • 4941.08 NOK / 100µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057568.1
PDZD11 / PDZK11 / PISP Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
51248
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PDZD11
Recombinant Protein
E. coli
MDSRIPYDDY PVVFLPAYEN PPAWIPPHER VHHPDYNNEL TQFLPRTITL KKPPGAQLGF NIRGGKASQL GIFISKVIPD SDAHRAGLQE GDQVLAVNDV DFQDIEHSKA VEILKTAREI SMRVRFFPYN YHRQKERTVH
Videos
SDS
Documents
Certificates
Special Offers

Special Offers