missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OXNAD1 Protein
A DNA sequence encoding the Homo sapiens (Human) OXNAD1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7897-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_612390.1 | |
OXNAD1 Protein | |
Research Use Only | |
OXNAD1 | |
Recombinant Protein | |
E. coli | |
AIRIEAASLR LTLSTLRHLT LTSIMKSKRK TDHMERTASV LRREIVSAAK VCGAASESPS VKSLRLLVAD QDFSFKAGQW VDFFIPGVSV VGGFSICSSP RLLEQERVIE LAVKYTNHPP ALWVHNTCTL DCEVAVRVGG EFFFDPQPAD ASRNLVLIAG GVGINPLLSI LRHAADLLRE QANKRNGYEI GTIKLFYSAK NTSELLFKKN ILDLVNEFPE KIACSLHVTK QTTQINAELK PYITEGRITE KEIRDHISKE TLFYICGPPP MTDFFSKQLE NNHVPKEHIC FEKWW | |
Greater than 90% as determined by SDS-PAGE. |
92106 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |