missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NRN1 / Neuritin 1 Protein

A DNA sequence encoding the Homo sapiens (Human) NRN1 / Neuritin 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8091-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002621

  • 22559.24 NOK / 1mg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265639.1
NRN1 / Neuritin 1 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
51299
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NRN1
Recombinant Protein
E. coli
AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGNG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers