Learn More
enQuireBio™ Recombinant Human Notchless protein homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Notchless protein homolog 1, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_001014445.1 |
---|---|
Gene ID (Entrez) | 54475 |
Name | Notchless protein homolog 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083391
|
enQuireBio™
QP8226-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003401
|
enQuireBio™
QP8226-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093391
|
enQuireBio™
QP8226-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013401
|
enQuireBio™
QP8226-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023401
|
enQuireBio™
QP8226-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033401
|
enQuireBio™
QP8226-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001014445.1 | |
Notchless protein homolog 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
54475 | |
Research Use Only | |
NLE1 | |
Recombinant Protein | |
E. coli | |
AAAVPDEAVA RDVQRLLVQF QDEGGQLLGS PFDVPVDITP DRLQLVCNAL LAQEDPLPLA FFVHDAEIVS SLGKTLESQA VETEKVLDII YQPQAIFRVR AVTRCTSSLE GHSEAVISVA FSPTGKYLAS GSGDTTVRFW DLSTETPHFT CKGHRHWVLS ISWSPDGRKL ASGCKNGQIL LWDPSTGKQV GRTLAGHSKW ITGLSWEPLH ANPECRYVAS SSKDGSVRIW DTTAGRCERI LTGHTQSVTC LRWGGDGLLY SASQDRTIKV WRAHDGVLCR TLQGHGHWVN TMALSTDYAL RTGAFEPAEA SVNPQDLQGS LQELKERALS RYNLVRGQGP ERLVSGSDDF TLFLWSPAED KKPLTRMTGH QALINQVLFS PDSRIVASAS FDKSIKLWDG RTGKYLASLR GHVAAVYQIA WSADSRLLVS GSSDSTLKVW DVKAQKLAMD LPGHADEVYA VDWSPDGQRV ASGGKDKCLR IWRR | |
Greater than 90% as determined by SDS-PAGE. |