missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nephrin Protein

A DNA sequence encoding the Homo sapiens (Human) Nephrin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6429-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013250

  • 22559.24 NOK / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004637.1
Nephrin Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
4868
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NPHS1
Recombinant Protein
E. coli
QLAIPASVPR GFWALPENLT VVEGASVELR CGVSTPGSAV QWAKDGLLLG PDPRIPGFPR YRLEGDPARG EFHLHIEACD LSDDAEYECQ VGRSEMGPEL VSPRVILSIL VPPKLLLLTP EAGTMVTWVA GQEYVVNCVS GDAKPAPDIT ILLSGQTISD ISANVNEGSQ QKLFTVEATA RVTPRSSDNR QLLVCEASSP ALEAPIKASF TVNVLFPPGP PVIEWPGLDE GHVRA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers